missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTH Antibody (BGN/1F8), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-65676C
This item is not returnable.
View return policy
Description
PTH Monoclonal specifically detects PTH in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| PTH | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PTH | |
| Synthetic peptide corresponding to amino acids 1-34 of mature PTH conjugated to a proprietary carrier molecule | |
| 0.1 mL | |
| Apoptosis, Biologically Active Proteins, Cancer | |
| 5741 | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| BGN/1F8 | |
| ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| Parathormone, Parathyrin, parathyroid hormone, parathyroid hormone 1, PTH1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| 2 x 10^7with 1-34 PTH, 5 x 10^8 with 1-84 PTH | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction