missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTH Antibody (PTH/911), DyLight 405 - N-terminal, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54433V
This item is not returnable.
View return policy
Description
PTH Monoclonal antibody specifically detects PTH in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| PTH | |
| Monoclonal | |
| DyLight 405 | |
| Parathormone, Parathyrin, parathyroid hormone, parathyroid hormone 1, PTH1 | |
| A synthetic peptide around amino acids 1-34 of human mature-PTH-polypeptide (exact sequence is proprietary) (Uniprot: P01270) | |
| 100 μg | |
| Apoptosis, BiologicAll speciesy Active Proteins, Cancer | |
| 5741 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| PTH/911 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction