missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCNY Mouse anti-Human, Clone: OTI12F10, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808174
This item is not returnable.
View return policy
Description
Cyclins, such as CCNY, control cell division cycles and regulate cyclin-dependent kinases.Specifications
| CCNY | |
| Monoclonal | |
| Unconjugated | |
| CCNY | |
| C10orf9, CBCP1, CCNX, CFP1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 219771 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI12F10 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8ND76 | |
| CCNY | |
| Full length human recombit protein of human CCNY produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction