missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTH, Mouse anti-Human, Clone: 3G9, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00005741-M11.100ug
This item is not returnable.
View return policy
Description
Sequence: SVSEIQLMHNLGKHLNSMERVEWLRKKLQDVHNFVALGAPLAPRDAGSQRPRKKEDNVLVESHEKSLGEADKADVNVLTKAKSQSpecifications
| PTH | |
| Monoclonal | |
| Unconjugated | |
| parathyroid hormone | |
| PTH1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 3G9 | |
| In 1x PBS, pH 7.4 | |
| N/A | |
| PTH | |
| PTH (NP_000306, 32 a.a.-115 a.a.) full-length recombinant protein. | |
| RUO | |
| 5741 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction