Learn More
Invitrogen™ PTH1R Recombinant Rabbit Monoclonal Antibody (003)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA530799
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human PTH1R/PTHR1.
Parathyroid hormone receptor 1 is a member of the G-protein coupled receptor family. This protein is a receptor for parathyroid hormone (PTH) and for parathyroid hormone-like hormone (PTHLH). The activity of this receptor is mediated by G proteins which activate adenylyl cyclase and also a phosphatidylinositol-calcium second messenger system. Defects in this receptor are known to be the cause of Jansen's metaphyseal chondrodysplasia (JMC), chondrodysplasia Blomstrand type (BOCD), as well as enchodromatosis. Parathyroid hormone receptor 1 has been reported to be expressed in bone, breast, brain, colon, GI, kidney, liver, placenta, skin, umbilical cord, and uterus.
Specifications
| PTH1R | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q03431 | |
| Pth1r | |
| Recombinant Human PTH1R/PTHR1 protein (Met1-Gly188). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA | |
| 003 | |
| Unconjugated | |
| Pth1r | |
| MGC138426; MGC138452; Parathormone; Parathyrin; parathyroid hormone 1 receptor; parathyroid hormone receptor; parathyroid hormone receptor 1; parathyroid hormone/parathyroid hormone-related peptide receptor; parathyroid hormone/parathyroid hormone-related protein receptor; PFE; PPR; PTH; PTH/PTHr receptor; PTH/PTHrP receptor; PTH/PTHrP type I receptor; PTH1 receptor; PTH1R; Pthr; Pthr1; PTHrel; PTH-related peptide receptor; PTHrP-R; PTHrP-receptor; seven transmembrane helix receptor | |
| Rabbit | |
| Protein A | |
| RUO | |
| 5745 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.