missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTH2R Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94871-0.02ml
This item is not returnable.
View return policy
Description
PTH2R Polyclonal antibody specifically detects PTH2R in Human samples. It is validated for Western Blot
Specifications
| PTH2R | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| parathyroid hormone 2 receptor, PTH2 receptor, PTHR2parathyroid hormone receptor 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 27-145 of human PTH2R (NP_005039.1). DSDGTITIEEQIVLVLKAKVQCELNITAQLQEGEGNCFPEWDGLICWPRGTVGKISAVPCPPYIYDFNHKGVAFRHCNPNGTWDFMHSLNKTWANYSDCLRFLQPDISIGKQEFFERLY | |
| 0.02 mL | |
| GPCR, Neuroscience, Signal Transduction | |
| 5746 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction