missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PTK7/CCK4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30221
This item is not returnable.
View return policy
Description
PTK7/CCK4 Polyclonal antibody specifically detects PTK7/CCK4 in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| PTK7/CCK4 | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| CCK-4, CCK4Colon carcinoma kinase 4, colon carcinoma kinase-4, Protein-tyrosine kinase 7, Pseudo tyrosine kinase receptor 7, PTK7 protein tyrosine kinase 7, Tyrosine-protein kinase-like 7 | |
| Between 700 and 750 | |
| 100 μg | |
| Protein Kinase | |
| 5754 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction