missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTK7/CCK4 Antibody (OTI2E7), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73708JF549
This item is not returnable.
View return policy
Description
PTK7/CCK4 Monoclonal specifically detects PTK7/CCK4 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| PTK7/CCK4 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PTK7 | |
| Full length recombinant protein of human PTK7/CCK4 (NP_690619) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| OTI2E7 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| CCK-4, CCK4Colon carcinoma kinase 4, colon carcinoma kinase-4, Protein-tyrosine kinase 7, Pseudo tyrosine kinase receptor 7, PTK7 protein tyrosine kinase 7, Tyrosine-protein kinase-like 7 | |
| Mouse | |
| Affinity Purified | |
| Protein Kinase | |
| 5754.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction