missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTK7 Monoclonal Antibody (OTI1G4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504077
This item is not returnable.
View return policy
Description
Receptor protein tyrosine kinases transduce extracellular signals across the cell membrane. A subgroup of these kinases lack detectable catalytic tyrosine kinase activity but retain roles in signal transduction. The protein encoded by this gene is a member of this subgroup of tyrosine kinases and may function as a cell adhesion molecule. This gene is thought to be expressed in colon carcinomas but not in normal colon, and therefore may be a marker for or may be involved in tumor progression. Four transcript variants encoding four different isoforms have been found for this gene.Specifications
| PTK7 | |
| Monoclonal | |
| 0.85 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q13308 | |
| PTK7 | |
| Full length human recombit protein of human PTK7 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| OTI1G4 | |
| Unconjugated | |
| PTK7 | |
| CCK-4, CCK4 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5754 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction