missing translation for 'onlineSavingsMsg'
Learn More

PTK7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. p-7208159 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16817784 20 μL 20µL
16807784 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16817784 Supplier Proteintech Supplier No. 177991AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The function remains unknown.

Specifications

Antigen PTK7
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.36 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene PTK7
Gene Accession No. Q13308, Q8BKG3
Gene Alias CCK 4, CCK4, Colon carcinoma kinase 4, Protein tyrosine kinase 7, PTK7, PTK7 protein tyrosine kinase 7, Tyrosine protein kinase like 7
Gene Symbols Ptk7
Host Species Rabbit
Immunogen PTK7 Fusion Protein Ag12004
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 301242, 5754, 71461
Target Species Human, Rat, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.