missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTOV1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 196.00 - € 468.00
Specifications
| Antigen | PTOV1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
PTOV1 Polyclonal antibody specifically detects PTOV1 in Human samples. It is validated for Western BlotSpecifications
| PTOV1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Biology, Cell Cycle and Replication | |
| PBS (pH 7.3), 50% glycerol | |
| 53635 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| ACID2, Activator interaction domain-containing protein 2, DKFZp586I111, MGC71475, prostate tumor overexpressed 1, prostate tumor overexpressed gene 1, PTOV-1 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human PTOV1 (NP_001292037.1). MRSPAVPTPARGQLGVAFVLLPPHSEGARVFGALGPIGPSSPGLTLGGLAVSEHRLSNKLLAWSGVLEWQEKRRPYSDSTAKLKRTLPCQAYVNQGENLE | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title