missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTOV1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-68998
This item is not returnable.
View return policy
Description
PTOV1 Polyclonal antibody specifically detects PTOV1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| PTOV1 | |
| Polyclonal | |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| ACID2, Activator interaction domain-containing protein 2, DKFZp586I111, MGC71475, prostate tumor overexpressed 1, prostate tumor overexpressed gene 1, PTOV-1 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: WSGVMEWQEPRPEPNSRSKRWLPSHVYVNQGEILRT | |
| 100 μg | |
| Cancer, Cell Biology, Cell Cycle and Replication | |
| 53635 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction