missing translation for 'onlineSavingsMsg'
Learn More

PTP epsilon Antibody [DyLight 488], Novus Biologicals Biologicals™

Product Code. 30511822 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30511822 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30511822 Supplier Novus Biologicals Supplier No. NBP338398G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PTP epsilon Polyclonal antibody specifically detects PTP epsilon in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTP epsilon
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias DKFZp313F1310, EC 3.1.3.48, FLJ57799, FLJ58245, HPTPE, protein tyrosine phosphatase epsilon, protein tyrosine phosphatase, receptor type, E, protein tyrosine phosphatase, receptor type, epsilon polypeptide, Protein-tyrosine phosphatase epsilon, PTPE, receptor-type tyrosine-protein phosphatase epsilon, R-PTP-EPSILON
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 20-200 of human PTP epsilon (NP_006495.1).,, Sequence:, LRGNETTADSNETTTTSGPPDPGASQPLLAWLLLPLLLLLLVLLLAAYFFRFRKQRKAVVSTSDKKMPNGILEEQEQQRVMLLSRSPSGPKKYFPIPVEHLEEEIRIRSADDCKQFREEFNSLPSGHIQGTFELANKEENREKNRYPNILPNDHSRVILSQLDGIPCSDYINASYIDGYKE
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Phosphatase, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 5791
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.