missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTP epsilon Antibody (OTI5F2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-02374
This item is not returnable.
View return policy
Description
PTP epsilon Monoclonal specifically detects PTP epsilon in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| PTP epsilon | |
| Monoclonal | |
| 0.58 mg/mL | |
| Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:50 | |
| PTPRE | |
| Full length human recombinant protein of human PTPRE (NP_006495) produced in HEK293T cell. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| OTI5F2 | |
| Unconjugated | |
| DKFZp313F1310, EC 3.1.3.48, FLJ57799, FLJ58245, HPTPE, protein tyrosine phosphatase epsilon, protein tyrosine phosphatase, receptor type, E, protein tyrosine phosphatase, receptor type, epsilon polypeptide, Protein-tyrosine phosphatase epsilon, PTPE, receptor-type tyrosine-protein phosphatase epsilon, R-PTP-EPSILON | |
| Mouse | |
| 80.6 kDa | |
| 0.1 mL | |
| GPCR, Protein Phosphatase, Tyrosine Kinases | |
| 5791 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction