missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PTP1B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582462
This item is not returnable.
View return policy
Description
Immunogen sequence: RFSYLAVIEG AKFIMGDSSV QDQWKELSHE DLEPPPEHIP PPPRPPKRIL EPHNGKCREF FPNHQWVKEE TQEDKDCPIK EEKGSPLNAA PYGIESMSQD TEVRSRVVGG SLRGAQAASP AKGEPSLPEK DEDHALSYW Highest antigen sequence indentity to the following orthologs: Rat - 67%, Mouse - 64%.
PTP1B (PTPN1) is a tyrosine-protein phosphatase which acts as a regulator of endoplasmic reticulum unfolded protein response. It mediates dephosphorylation of EIF2AK3/PERK and in turn inactivates the protein kinase activity of EIF2AK3/PERK. PTP1B also plays a role in CKII- and p60c-src-induced signal transduction cascades. It regulates the EFNA5-EPHA3 signaling pathway which modulates cell reorganization and cell-cell repulsion. PTP1B also regulates the hepatocyte growth factor receptor signaling pathway through dephosphorylation of MET.
Specifications
| PTP1B | |
| Polyclonal | |
| Unconjugated | |
| PTPN1 | |
| CPTP1; EC 3.1.3.48; EGK_02300; hgnc9642; KIAA0283; protein tyrosine phosphatase 1B; protein tyrosine phosphatase non-receptor type 1; protein tyrosine phosphatase, non-receptor type 1; protein tyrosine phosphatase, placental; protein-protein-tyrosine phosphatase HA2; protein-tyrosine phosphatase 1B; Protein-tyrosine phosphatase HA2; PTN1; Ptp; PTP1B; PTP-1B; ptp1b protein; PTP-HA2; Ptpn1; RP5-1121H13.2; RPTPrho; tyrosine phosphatase 1b; tyrosine-phosphatase; Tyrosine-protein phosphatase non-receptor type 1; wu:fk54h03 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5770 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P18031 | |
| PTPN1 | |
| Recombinant protein corresponding to Human PTPN1. Recombinant protein control fragment (Product #RP-90585). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction