missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTP1B/PTPN1 Antibody (226), mFluor Violet 610 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89400MFV610
This item is not returnable.
View return policy
Description
PTP1B/PTPN1 Monoclonal antibody specifically detects PTP1B/PTPN1 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
Specifications
| PTP1B/PTPN1 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunoprecipitation | |
| 226 | |
| Western Blot, ELISA, Immunoprecipitation | |
| EC 3.1.3.48, protein tyrosine phosphatase 1B, protein tyrosine phosphatase, non-receptor type 1, Protein-tyrosine phosphatase 1B, PTP-1B, PTP1Bprotein tyrosine phosphatase, placental, tyrosine-protein phosphatase non-receptor type 1 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human PTP1B/PTPN1 (Accession#: NP_002818.1; Glu 2-Asn 321). | |
| 0.1 mL | |
| Phospho Specific | |
| 5770 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction