missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTP1B/PTPN1 Antibody (OTI1D10), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73729V
This item is not returnable.
View return policy
Description
PTP1B/PTPN1 Monoclonal specifically detects PTP1B/PTPN1 in Human, Rat, Canine, Monkey, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated.
Specifications
| PTP1B/PTPN1 | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PTPN1 | |
| Full length recombinant protein of human PTP1B/PTPN1 (NP_002818) produced in HEK293 cell. | |
| 0.1 mL | |
| Primary | |
| Human, Rat, Canine, Monkey, Primate | |
| IgG2a |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay | |
| OTI1D10 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated | |
| EC 3.1.3.48, protein tyrosine phosphatase 1B, protein tyrosine phosphatase, non-receptor type 1, Protein-tyrosine phosphatase 1B, PTP-1B, PTP1Bprotein tyrosine phosphatase, placental, tyrosine-protein phosphatase non-receptor type 1 | |
| Mouse | |
| Affinity Purified | |
| Phospho Specific | |
| 5770.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction