missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTP1B/PTPN1 Antibody (OTI1D10), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73729MFV500
This item is not returnable.
View return policy
Description
PTP1B/PTPN1 Monoclonal antibody specifically detects PTP1B/PTPN1 in Human, Rat, Canine, Monkey, Primate samples. It is validated for Flow Cytometry
Specifications
| PTP1B/PTPN1 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Rat, Canine, Monkey, Primate | |
| Purified |
| Flow Cytometry | |
| OTI1D10 | |
| Flow Cytometry | |
| EC 3.1.3.48, protein tyrosine phosphatase 1B, protein tyrosine phosphatase, non-receptor type 1, Protein-tyrosine phosphatase 1B, PTP-1B, PTP1Bprotein tyrosine phosphatase, placental, tyrosine-protein phosphatase non-receptor type 1 | |
| Full length human recombinant protein of human PTPN1(NP_002818) produced in HEK293 cell. | |
| 0.1 mL | |
| Phospho Specific | |
| 5770 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction