missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PTPLB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113050
This item is not returnable.
View return policy
Description
PTPLB Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
PTPLB is probable anti-phosphatase.
Specifications
| PTPLB | |
| Polyclonal | |
| Unconjugated | |
| Hacd2 | |
| 3-hydroxyacyl-CoA dehydratase 2; 6330408J20Rik; AI255777; AI481689; HACD2; Hcad2; I79_000662; protein tyrosine phosphatase-like (proline instead of catalytic arginine), member b; protein-tyrosine phosphatase-like member B; Ptplb; very-long-chain (3R)-3-hydroxyacyl-[acyl-carrier protein] dehydratase 2; very-long-chain (3R)-3-hydroxyacyl-CoA dehydratase 2 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 201562 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q6Y1H2 | |
| Hacd2 | |
| Recombinant Human Very-long-chain (3R)-3-hydroxyacyl-CoA dehydratase 2 protein (7-41AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction