missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PTPN12 Monoclonal Antibody (AG25)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA112377
This item is not returnable.
View return policy
Description
A suggested positive control for this product is SK-N-SH cell lysate.
PTP PEST is an 88 kDa cytosolic protein tyrosine phosphatase which is ubiquitiously expressed in mammalian tissues. PTP PEST exhibits a high specific activity when assayed in vitro with artificial tyrosine-phosphorylated substrates. PTP PEST is subject to regulation via phosphorylation of ser 39 and is catalyzed by both protein kinase C and protein kinase A.
Specifications
| PTPN12 | |
| Monoclonal | |
| Unconjugated | |
| PTPN12 | |
| MPTP-PEST; P19-PTP; pest; protein tyrosine phosphatase non-receptor type 12; protein tyrosine phosphatase, non-receptor type 12; protein-tyrosine phosphatase G1; Protein-tyrosine phosphatase P19; PTPG1; PTPN12; PTP-P19; PTP-PEST; RKPTP; tcag7.1075; Tyrosine-protein phosphatase non-receptor type 12 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 19248, 5782 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| AG25 | |
| PBS with 0.08% sodium azide | |
| P35831, Q05209 | |
| PTPN12 | |
| Baculovirus-expressed human full-length PTP-PEST. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction