missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PTPN20 Recombinant Protein Antigen
Shop All Bio Techne Products
Click to view available options
Protein Length:
MALELKNLPGEFNSGNQPSNREKNRYRDILP
NFPHNDYEDVFEEPSESGSDPSMWTARG
Unit Size:
0.10mL
0.1mL
Description
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human PTPN20 The PTPN20 Recombinant Protein Antigen is derived from E. coli. The PTPN20 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
Specifications
Specifications
| Protein Length | MALELKNLPGEFNSGNQPSNREKNRYRDILP |
| Purity | >80% by SDS-PAGE and Coomassie blue staining |
| Common Name | PTPN20 |
| Content And Storage | Store at −20°C short term. Avoid freeze-thaw cycles. |
| Formulation | PBS and 1M Urea, pH 7.4 |
| For Use With (Application) | AC |
| Gene Alias | protein tyrosine phosphatase, non-receptor type 20, bA42B19.1, protein tyrosine phosphatase, non-receptor type 20B |
| Gene Symbol | PTPN20 |
| Label Type | Unlabeled |
| Molecular Weight (g/mol) | 21 kDa |
| Show More |
For Research Use Only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction