missing translation for 'onlineSavingsMsg'
Learn More

PTPN3 Antibody - BSA Free, Novus Biologicals™

Product Code. 18637031 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.01mL
0.02mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18637031 0.02 mL 0.02mL
18651651 0.1 mL 0.01mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18637031 Supplier Novus Biologicals Supplier No. NBP2939450.02ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PTPN3 Polyclonal antibody specifically detects PTPN3 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTPN3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000
Formulation PBS (pH 7.3), 50% glycerol
Gene Alias DKFZp686N0569, EC 3.1.3.48, protein tyrosin phosphatase H1, protein tyrosine phosphatase, non-receptor type 3, Protein-tyrosine phosphatase H1, PTP-H1, PTPH1cytoskeletal-associated protein tyrosine phosphatase, tyrosine phosphatase, tyrosine-protein phosphatase non-receptor type 3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human PTPN3 (NP_001138840.1). MTSRLRALGGRINNIRTSELPKEKTRSEVICSIHFLDGVVQTFKVTKQDTGQVLLDMVHNHLGVTEKEYFGLQHDDDSVDSPRWLEASKAIRKQLKGGFPCTLHFRVRFFIPDPNTLQQEQTRHLYFLQLKMDICEGRLTCPLNSAVVLASYAVQSHFGDYNSSIHHPGYLSDSHFIPDQNEDFLTKVESLHEQHSGLKQSEAESCYINIARTLDFYGVELHSGRDLHNLDLMIGIASAGVAVYRKYICTSFYPWVNILKISFKRKKFFIHQRQKQAESREHIVAFNMLNYRSCKNLWKS
Purification Method Affinity purified
Quantity 0.02 mL
Regulatory Status RUO
Research Discipline Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5774
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.