missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTPN7 Antibody (OTI3B10), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73732AF750
This item is not returnable.
View return policy
Description
PTPN7 Monoclonal specifically detects PTPN7 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| PTPN7 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| PTPN7 | |
| Full length recombinant protein of human PTPN7 (NP_542155) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine | |
| IgG2b |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| OTI3B10 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| BPTP-4, FLJ18703, HEPTPnonreceptor-type, stress induced, LPTP, protein tyrosine phosphatase, non-receptor type 7, tyrosine-protein phosphatase non-receptor type 7 | |
| Mouse | |
| Affinity Purified | |
| Cancer | |
| 5778.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction