missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ptprc, Rat anti-Mouse, CF-Blue, Clone: 30-F11, Abnova™
Rat Monoclonal Antibody
Brand: Abnova MAB15485.100ug
This item is not returnable.
View return policy
Specifications
| Ptprc | |
| Monoclonal | |
| CF-Blue | |
| In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide). | |
| B220/CD45R/Cd45/Ly-5/Lyt-4/T200/loc | |
| Rat | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2b |
| Flow Cytometry | |
| 30-F11 | |
| Flow Cytometry The optimal working dilution should be determined by the end user. | |
| protein tyrosine phosphatase, receptor type, C | |
| Ptprc | |
| Mouse thymocytes and splenocytes. | |
| RUO | |
| 19264 | |
| Store in the dark at 4°C. Avoid prolonged exposure to light. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction