missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTPRCAP Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-36565R
This item is not returnable.
View return policy
Description
PTPRCAP Polyclonal antibody specifically detects PTPRCAP in Human samples. It is validated for Western Blot
Specifications
| PTPRCAP | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Immunogen affinity purified | |
| Human | |
| Purified |
| Western Blot | |
| Western Blot | |
| CD45-APCD45 associated protein, CD45-associated protein, LPAPMGC138603, Lymphocyte phosphatase-associated phosphoprotein, MGC138602, protein tyrosine phosphatase receptor type C-associated protein, protein tyrosine phosphatase, receptor type, c polypeptide-associated protein, protein tyrosine phosphatase, receptor type, C-associated protein, PTPRC-associated protein | |
| 0.1 mL | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction