Learn More
Invitrogen™ PTPRD Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA590641
Description
Immunogen sequence: TAKTAPDVLR TKPAFIGKTN LDGMITVQLP EVPANENIKG YYIIIVPLKK SRGKFIKPWE SPDEMELDEL LKEISRKRRS IRYGREVELK PYIAAHFDVL PTEFTLGDDK HYGGFTNKQL QSGQEYVFFV LAVMEHAESK MYATSPYSDP VVSMDLDPQP ITDEEE; Positive Samples: 293T, OVCAR3, Raji, Mouse brain, Mouse kidnay, Mouse heart, Rat brain; Cellular Location: Membrane, Single-pass type I membrane protein.
PTPRD is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP contains an extracellular region, a single transmembrane segment and two tandem intracytoplasmic catalytic domains, and thus represents a receptor-type PTP. The extracellular region of this protein is composed of three Ig-like and eight fibronectin type III-like domains. Studies of the similar genes in chicken and fly suggest the role of this PTP is in promoting neurite growth, and regulating neurons axon guidance.
Specifications
| PTPRD | |
| Polyclonal | |
| Unconjugated | |
| PTPRD | |
| 1110002J03Rik; 3000002J10Rik; B230219D21Rik; HPTP; HPTPD; HPTPDELTA; protein tyrosine phosphatase, receptor type D; protein tyrosine phosphatase, receptor type, D; protein tyrosine phosphatase, receptor type, delta polypeptide; Protein-tyrosine phosphatase delta; PTPD; Ptprd; receptor-type tyrosine-protein phosphatase delta; RPTPDELTA; R-PTP-delta | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 19266, 313278, 5789 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.64 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| P23468, Q64487 | |
| PTPRD | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1100-1265 of human PTPRD (NP_0028301). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.