missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTPRE Monoclonal Antibody (OTI3B8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501027S
This item is not returnable.
View return policy
Description
PTPRE Monoclonal antibody specifically detects PTPRE in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| PTPRE | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P23469 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 5791 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Western Blot | |
| OTI3B8 | |
| Unconjugated | |
| PTPRE | |
| HPTPE; protein tyrosine phosphatase epsilon; protein tyrosine phosphatase epsilon-like 1; protein tyrosine phosphatase epsilon-like 2; protein tyrosine phosphatase, receptor type E; protein tyrosine phosphatase, receptor type, E; protein tyrosine phosphatase, receptor type, epsilon; protein tyrosine phosphatase, receptor type, epsilon polypeptide; protein-tyrosine phosphatase epsilon; PTP epsilon; PTPE; PTPepsilon; PTPRE; PTPvarepsilon; receptor-type tyrosine-protein phosphatase epsilon; RPTPepsilon; R-PTP-epsilon | |
| Full length human recombinant protein of human PTPRE produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction