missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PTPRE Monoclonal Antibody (OTI3D4)
GREENER_CHOICE

Product Code. 15974674
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15974674 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15974674 Supplier Invitrogen™ Supplier No. MA525072

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PTPRE Monoclonal Antibody for Flow, ICC/IF, IHC (P), Western Blot

The protein encoded by this gene is a member of the protein tyrosine phosphatase family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. Two alternatively spliced transcript variants of this gene have been reported, one of which encodes a receptor-type PTP that possesses a short extracellular domain, a single transmembrane region, and two tandem intracytoplasmic catalytic domains; Another one encodes a PTP that contains a distinct hydrophilic N-terminus, and thus represents a nonreceptor-type isoform of this PTP. Studies of the similar gene in mice suggested the regulatory roles of this PTP in RAS related signal transduction pathways, cytokines induced SATA signaling, as well as the activation of voltage-gated K+ channels.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTPRE
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI3D4
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PTPRE
Gene Accession No. P23469
Gene Alias HPTPE; protein tyrosine phosphatase epsilon; protein tyrosine phosphatase epsilon-like 1; protein tyrosine phosphatase epsilon-like 2; protein tyrosine phosphatase, receptor type E; protein tyrosine phosphatase, receptor type, E; protein tyrosine phosphatase, receptor type, epsilon; protein tyrosine phosphatase, receptor type, epsilon polypeptide; protein-tyrosine phosphatase epsilon; PTP epsilon; PTPE; PTPepsilon; PTPRE; PTPvarepsilon; receptor-type tyrosine-protein phosphatase epsilon; RPTPepsilon; R-PTP-epsilon
Gene Symbols PTPRE
Host Species Mouse
Immunogen Full length human recombinant protein of PTPRE produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5791
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.