Learn More
Invitrogen™ PTPRF Monoclonal Antibody (N165/38), FITC
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA545638
Product Code. 17827032
  • € 467.00 / 100µg
Description
97% identical in both rat and mouse. >80% identity with PTPRD and PTPRS. >50% identity with PTPRM and PTPRK. 1 μg/mL of MA5-45638 was sufficient for detection of LAR/PTPRF in 20 μg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 85kDa (full length protein is 210 kDa - smaller due to proteolysis into P-subunit containing transmembrane and intracellular domains. This antibody was formerly sold as clone S165-38.
Possible cell adhesion receptor. It possesses an intrinsic protein tyrosine phosphatase activity (PTPase).The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PTPRF
Monoclonal
1 mg/mL
PBS, 9.1mM sodium bicarbonate with 640.91mM DMSO, 136.36mM ethanolamine and no preservative; pH 7.4
A2A8L5, P10586, Q64604
PTPRF
Fusion protein amino acids 1315-1607 (cytoplasmic C-terminus) of human LAR.
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG2a
Western Blot, Immunocytochemistry
N165/38
FITC
PTPRF
AA591035; BNAH2; FLJ43335; FLJ45062; FLJ45567; LAR; LAR protein; LARFN5C; LARS; LCA homolog; LCA-homolog; leukocyte antigen-related (LAR) PTP receptor; leukocyte antigen-related tyrosine phosphatase; leukocyte common antigen related; protein tyrosine phosphatase receptor-type F; protein tyrosine phosphatase, receptor type F; protein tyrosine phosphatase, receptor type, F; protein tyrosine phosphatase, receptor type, F polypeptide; Ptprf; PTPRF protein; receptor-linked protein-tyrosine phosphatase LAR; receptor-type tyrosine-protein phosphatase F; RPTP-LAR
Mouse
Protein G
RUO
19268, 360406, 5792
4°C, store in dark