missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PTPRH Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

€ 156.00 - € 441.00

Specifications

Antigen PTPRH
Concentration 0.5 mg/mL
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17710784
GREENER_CHOICE
View Documents
Invitrogen™
PA5119375
200 μL
€ 441.00
200µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17703064
View Documents
Invitrogen™
PA5121662
20 μL
€ 156.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PTPRH Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP possesses an extracellular region, a single transmembrane region, and a single intracytoplasmic catalytic domain, and thus represents a receptor-type PTP. The extracellular region contains eight fibronectin type III-like repeats and multiple N-glycosylation sites. The gene was shown to be expressed primarily in brain and liver, and at a lower level in heart and stomach. It was also found to be expressed in several cancer cell lines, but not in the corresponding normal tissues.
Specifications

Specifications

PTPRH
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Polyclonal
Liquid
IgG
Mouse
PTPRH
Bem2; brain-enriched membrane-associated protein tyrosine BEM-2; protein tyrosine phosphatase, receptor type H; protein tyrosine phosphatase, receptor type, H; PTPRH; receptor-type tyrosine-protein phosphatase H; R-PTP-H; SAP1; SAP-1; Stomach cancer-associated protein tyrosine phosphatase 1; transmembrane-type protein-tyrosine phosphatase type H
PTPRH
Primary
Antigen affinity chromatography, Protein A
0.5 mg/mL
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
E9Q0N2
545902
Recombinant protein Tyrosine Phosphatase Receptor Type H. The antigen corresponds to amino acid range 685-909 of the target protein.
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.