missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PTPRM Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110675
This item is not returnable.
View return policy
Description
Immunogen sequence: STRQEMTVMV NSMDKSYAEQ GTNCDEAFSF MDTHNLNGRS VSSPSSFTMK TNTLSTSVPN SYYPDETHTM ASDTSSLVQS HTYKKREPAD VPYQTGQLHP AIRVADLLQH ITQMKCAEGY GFKEEYEVST Highest antigen sequence indentity to the following orthologs: Rat - 89%, Mouse - 95%.
The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP possesses an extracellular region, a single transmembrane region, and two tandem catalytic domains, and thus represents a receptor-type PTP. The extracellular region contains a meprin-A5 antigen-PTP mu (MAM) domain, an Ig-like domain and four fibronectin type III-like repeats. This PTP has been shown to mediate cell-cell aggregation through the interaction with another molecule of this PTP on an adjacent cell. This PTP can interact with scaffolding protein RACK1/GNB2L1, which may be necessary for the downstream signaling in response to cell-cell adhesion. Alternative splicing results in multiple transcripts encoding distinct isoforms.
Specifications
| PTPRM | |
| Polyclonal | |
| Unconjugated | |
| PTPRM | |
| hR-PTPu; Kiaa4044; MGC166994; mKIAA4044; protein tyrosine phosphatase mu; protein tyrosine phosphatase receptor type M; protein tyrosine phosphatase receptor-type M; protein tyrosine phosphatase, receptor type M; protein tyrosine phosphatase, receptor type, M; protein tyrosine phosphatase, receptor type, mu polypeptide; protein tyrosine phosphatase, receptor-type, M; protein-tyrosine phosphatase mu; PTP mu; Ptpmu; PTPRL1; PTPRM; receptor-type tyrosine-protein phosphatase mu; RPTPM; RPTPmu; R-PTP-MU; RPTPU | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5797 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.20 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P28827 | |
| PTPRM | |
| Recombinant Protein corresonding to Human PTPRM. Recombinant protein control fragment (Product #RP-105695). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction