missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTPRN2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-81629-25ul
This item is not returnable.
View return policy
Description
PTPRN2 Polyclonal antibody specifically detects PTPRN2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| PTPRN2 | |
| Polyclonal | |
| Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| EC 3.1.3.48, IA-2beta, IAR/receptor-like protein-tyrosine phosphatase, ICAARIARPTPRP, Islet cell autoantigen-related protein, KIAA0387, phogrin, protein tyrosine phosphatase receptor pi, protein tyrosine phosphatase, receptor type, N polypeptide 2, receptor-type tyrosine-protein phosphatase N2, R-PTP-N2, tyrosine phosphatase IA-2 beta | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: RFSESILTYVAHTSALTYPPGSRTQLREDLLPRTLGQLQPDELSPKVDSGVDRHHLMAALSAYAAQRPPAPPGEGSLEPQYLLRAPSRMPRPLLAPAAPQKWPSPLGDSEDPSST | |
| 25 μL | |
| Lipid and Metabolism, Protein Phosphatase | |
| 5799 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction