missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTPRN2 Monoclonal Antibody (8E12)

PTPRN2 Monoclonal Antibody (8E12)
Brand: Invitrogen MA519759
This item is not returnable.
View return policy
Description
Specifications
| PTPRN2 | |
| Monoclonal | |
| 0.4 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q92932 | |
| PTPRN2 | |
| PTPRN2 (NP_002838, 108 a.a. ~ 212 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Western Blot | |
| 8E12 | |
| Unconjugated | |
| PTPRN2 | |
| 4930425H11Rik; EC 3.1.3.48; IA-2 beta; IA2beta; IA-2beta; IA-2beta60; IA-2beta64; IA-2beta71; IAR; IAR/receptor-like protein-tyrosine phosphatase; ICAAR; Islet cell autoantigen-related protein; KIAA0387; mKIAA0387; phogrin; Phol; protein tyrosine phosphatase receptor pi; protein tyrosine phosphatase receptor-type N polypeptide 2; protein tyrosine phosphatase, receptor type N2; protein tyrosine phosphatase, receptor type, N polypeptide 2; protein tyrosine phosphatase, receptor-type, N polypeptide 2; protein tyrosine phosphatase-NP; PTP IA-2beta; PTP NE-6; PTPNE6; PTP-NP; PTPRN2; PTPRP; Receptor-type tyrosine-protein phosphatase N2; R-PTP-N2; tyrosine phosphatase IA-2 beta | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 5799 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction