missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PTPRO Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556964
This item is not returnable.
View return policy
Description
Immunogen sequence: TRVNISYWEG KDFRTMLYKD FFKGKTVFNH WLPGMCYSNI TFQLVSEATF NKSTLVEYSG VSHEPKQHRT APYPPQNISV RIVNLNKNNW EEQSGNFPEE SFMRSQDTIG KEKLFHFTEE TPEIPSGNIS SGWP Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 84%.
Glomerular epithelial protein 1 (GLEPP1), also designated protein tyrosine phosphatase U2 (PTPase U2) or receptor-type tyrosine-protein phosphatase O (PTPRO), belongs to the protein-tyrosine phosphatase family. GLEPP1 is a Type I membrane protein containing 8 fibronectin type-III domains and 1 tyrosine-protein phosphatase domain. GLEPP1 is expressed primarily in the glomeruli of the kidney, but is also detected in placenta, lung and brain.
Specifications
| PTPRO | |
| Polyclonal | |
| Unconjugated | |
| PTPRO | |
| D28; FMI; GLEPP1; Glomerular epithelial protein 1; hPTP-J; NPHS6; osteoclastic transmembrane protein-tyrosine phosphatase; PCP2; PCP-2; phosphotyrosine phosphatase U2; protein tyrosine phosphatase PTP-U2; protein tyrosine phosphatase U2; protein tyrosine phosphatase, non-receptor type 15; protein tyrosine phosphatase, receptor type O; protein tyrosine phosphatase, receptor type, O; PTP phi; PTP pi; PTPase U2; PTPBK; PTP-J; Ptpn15; PTPoc; PTP-OC; PTPphi; Ptpro; PTP-RO; PTPROT; PTPRU; PTPU2; PTP-U2; receptor protein tyrosine phosphatase RO; receptor-type protein tyrosine phosphatase D30; receptor-type tyrosine-protein phosphatase O; RPTP-BK; R-PTP-O; R-PTP-psi; R-PTP-U | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5800 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q16827 | |
| PTPRO | |
| Recombinant protein corresponding to Human PTPRO. Recombinant protein control fragment (Product #RP-101062). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction