Learn More
Invitrogen™ PTPRS Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597164
Description
Immunogen sequence: TNRGSSLGGL QQTVTAWTAF NLLNGKPSVA PKPDADGFIM VYLPDGQSPV PVQSYFIVMV PLRKSRGGQF LTPLGSPEDM DLEELIQDIS RLQRRSLRHS RQLEVPRPYI AARFSVLPPT FHPGDQKQYG GFDNRGLEPG HRYVLFVLAV LQKSEPTFAA SPFSDPFQLD NPDPQPIVDG E; Positive Samples: Rat testis; Cellular Location: Membrane, Single-pass type I membrane protein.
PTPRS cell surface receptor that binds to glycosaminoglycans, including chondroitin sulfate proteoglycans and heparan sulfate proteoglycan. Binding to chondroitin sulfate and heparan sulfate proteoglycans has opposite effects on PTPRS oligomerization and regulation of neurite outgrowth. Contributes to the inhibition of neurite and axonal outgrowth by chondroitin sulfate proteoglycans, also after nerve transection. Plays a role in stimulating neurite outgrowth in response to the heparan sulfate proteoglycan GPC2. Required for normal brain development, especially for normal development of the pituitary gland and the olfactory bulb. Functions as tyrosine phosphatase. Mediates dephosphorylation of NTRK1, NTRK2 and NTRK3. Plays a role in down-regulation of signaling cascades that lead to the activation of Akt and MAP kinases. Down-regulates TLR9-mediated activation of NF-kappa-B, as well as production of TNF, interferon alpha and interferon beta.
Specifications
| PTPRS | |
| Polyclonal | |
| Unconjugated | |
| Ptprs | |
| AL022616; Larptp2; LAR-PTP2; Leukocyte common antigen-related protein-tyrosine phosphatase 2; leukocyte common antigen-related proten-tyrosine phosphatase 2; protein tyrosine phosphatase PTPsigma; protein tyrosine phosphatase, receptor type S; protein tyrosine phosphatase, receptor type, D; protein tyrosine phosphatase, receptor type, S; PTP; PTP-NU3; PTPNU-3; Ptprd; Ptprs; PTPsigma; Ptpsigma.; Ptpt9; Receptor-type tyrosine-protein phosphatase S; Receptor-type tyrosine-protein phosphatase sigma; Rptpd; R-PTP-S; RPTPsigma; R-PTP-sigma; tissue-specific protein tyrosine phosphatase | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 25529 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.06 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q64605 | |
| Ptprs | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1100-1280 of human PTPRS (NP_0028413). | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.