missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PTPRZ Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101832
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total PTPRZ1.
Protein tyrosine phosphatase that negatively regulates oligodendrocyte precursor proliferation in the embryonic spinal cord. Required for normal differentiation of the precursor cells into mature, fully myelinating oligodendrocytes. May play a role in protecting oligondendrocytes against apoptosis. May play a role in the establishment of contextual memory, probably via the dephosphorylation of proteins that are part of important signaling cascades.
Specifications
| PTPRZ | |
| Polyclonal | |
| Unconjugated | |
| PTPRZ1 | |
| 3F8 chondroitin sulfate proteoglycan; 3H1 keratan sulfate proteoglycan; 6B4 proteoglycan; AI850339; DSD-1-PG; HPTPZ; HPTPzeta; HTPZP2; phosphacan; protein tyrosine phosphatase, receptor type Z, polypeptide 1; protein tyrosine phosphatase, receptor type Z1; protein tyrosine phosphatase, receptor-type, Z polypeptide 1; protein tyrosine phosphatase, receptor-type, zeta polypeptide 1; protein tyrosine phosphate, receptor type, Z; Protein-tyrosine phosphatase receptor type Z polypeptide 1; Protein-tyrosine phosphatase receptor type Z polypeptide 2; PTP zeta; PTP18; PTPbeta; PTPRZ; PTPRZ1; PTPRZ2; Ptpz; PTPzeta; PTP-ZETA; PTPzeta-A; PTPzeta-B; PTPzeta-S; receptor-type tyrosine phosphatase beta/zeta; receptor-type tyrosine-protein phosphatase zeta; RPTPB; RPTPbeta; RPTPz; R-PTP-zeta; R-PTP-zeta-2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 19283, 25613, 5803 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| B9EKR1, P23471, Q62656 | |
| PTPRZ1 | |
| A synthesized peptide derived from human PTPRZ1(Accession P23471), corresponding to amino acid residues F121-K171. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction