missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTRH2 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-55963R
This item is not returnable.
View return policy
Description
PTRH2 Polyclonal specifically detects PTRH2 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| PTRH2 | |
| Polyclonal | |
| Western Blot | |
| Bcl-2 inhibitor of transcription 1, BIT1CGI-147, EC 3.1.1.29, mitochondrial, peptidyl-tRNA hydrolase 2, PTH2FLJ32471 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Bit1 (IN) | |
| Store at 4°C in the dark. |
| Western Blot | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PTRH2 | |
| Antibody was raised against a 16 amino acid synthetic peptide from near the center of human PTRH2. The immunogen is located within amino acids 80 - 130 of PTRH2. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction