missing translation for 'onlineSavingsMsg'
Learn More

PTRHD1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

€ 391.65 - € 540.75

Specifications

Antigen PTRHD1
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18453031
View Documents
Novus Biologicals
NBP1-93543-25ul
25 μL
ONLINE EXCLUSIVE PRICE
€ 415.00
€ 391.65 / 25µL
Save € 23.35
5% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18056486
View Documents
Novus Biologicals
NBP1-93543
0.1 mL
ONLINE EXCLUSIVE PRICE
€ 572.00
€ 540.75 / 0.10mL
Save € 31.25
5% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PTRHD1 Polyclonal specifically detects PTRHD1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

PTRHD1
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human
C2orf79, peptidyl-tRNA hydrolase domain containing 1, peptidyl-tRNA hydrolase domain-containing protein 1
PTRHD1
IgG
Affinity Purified
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200
Polyclonal
Rabbit
DNA replication Transcription Translation and Splicing
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
391356
This antibody was developed against Recombinant Protein corresponding to amino acids:GRMRKVVLEAPDETTLKELAETLQQKNIDHMLWLEQPENIATCIALRPYPKEEVGQYLKKFRLFK
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.