Learn More
PTS Antibody (02), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-06548-50ul
 View more versions of this product
Product Code. 18620368
  • € 220.00 / 50µL
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
PTS Monoclonal antibody specifically detects PTS in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Spécification
Spécification
PTS
Monoclonal
Unconjugated
0.2 um filtered solution in PBS
Mouse
Protein A purified
RUO
5805
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1
ELISA
02
ELISA 1:1000-1:2000
6-pyruvoyl tetrahydrobiopterin synthase, 6-pyruvoyltetrahydropterin synthase, EC 4.2.3.12, FLJ97081, PTPSPTP synthase
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human PTS (Uniprot#: Q03393; Met1-Glu145).