missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PU.1 Recombinant Rabbit Monoclonal Antibody (PD00-14)
GREENER_CHOICE

Product Code. 17949282
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17949282 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17949282 Supplier Invitrogen™ Supplier No. MA550702

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive control: Daudi cell lysate, THP-1 cell lysate, human B-cell lymphoma tissue. Predicted band size: 31 kDa Subcellular Location: Nucleus.

This gene encodes an ETS-domain transcription factor that activates gene expression during myeloid and B-lymphoid cell development. The nuclear protein binds to a purine-rich sequence known as the PU-box found near the promoters of target genes, and regulates their expression in coordination with other transcription factors and cofactors. The protein can also regulate alternative splicing of target genes. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PU.1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone PD00-14
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene SPI1
Gene Accession No. P17947
Gene Alias 31 kDa transforming protein; 31 kDa-transforming protein; 31kDa Transforming Protein; Dis1; Dis-1; hCG_25181; hematopoietic transcription factor PU.1; OF; Pu.1; SFFV proviral integration 1; SFFV proviral integration 1 protein; Sfpi1; Sfpi-1; Spi1; SPI-1; Spi-1 proto-oncogene; SPIA; SPI-A; spleen focus forming virus (SFFV) proviral integration oncogene; spleen focus forming virus (SFFV) proviral integration oncogene spi1; spleen focus forming virus proviral integration oncogene spi1; Tcfpu1; Tfpu.1; Transcription factor PU.1; Transcription Factor spi1
Gene Symbols SPI1
Host Species Rabbit
Immunogen Synthetic peptide within Human PU.1 aa 1-100.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6688
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.