missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PUF-A,KIAA0020 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | PUF-A,KIAA0020 |
|---|---|
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17541346
|
Proteintech
25491-1-AP-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17531346
|
Proteintech
25491-1-AP-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The PUF-A,KIAA0020 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human PUF-A,KIAA0020. This antibody recognizes human antigen. The PUF-A,KIAA0020 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| PUF-A,KIAA0020 | |
| Polyclonal | |
| Rabbit | |
| 9933 | |
| Primary |
| Western Blot, ELISA | |
| Unconjugated | |
| Human | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title