missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PUM1 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97707R
This item is not returnable.
View return policy
Description
PUM1 Polyclonal specifically detects PUM1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| PUM1 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| HSPUM, KIAA0099PUML1, PUMH, PUMH1pumilio homolog 1, pumilio (Drosophila) homolog 1, pumilio homolog 1 (Drosophila), Pumilio-1 | |
| Produced in rabbits immunized with E. coli-derived Human PUM1 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunofluorescence | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 9698 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction