missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PUM1 Antibody, FITC, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-259F
This item is not returnable.
View return policy
Description
PUM1 Polyclonal antibody specifically detects PUM1 in Human, Mouse samples. It is validated for Immunofluorescence
Specifications
| PUM1 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| HSPUM, KIAA0099PUML1, PUMH, PUMH1pumilio homolog 1, pumilio (Drosophila) homolog 1, pumilio homolog 1 (Drosophila), Pumilio-1 | |
| The immunogen recognized by this antibody maps to a region between residues 225 and 275 of human Pumilio 1 using the numbering given in entry NP_055491.1 (GeneID 9698). | |
| 0.1 mL | |
| Cell Biology, Stem Cells | |
| 9698 | |
| Store at 4C in the dark. | |
| IgG |
| Immunofluorescence | |
| FITC | |
| PBS | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction