Learn More
Invitrogen™ PUM2 Recombinant Rabbit Monoclonal Antibody (24GB1250)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557656
This item is not returnable.
View return policy
Description
PUM2 Recombinant Monoclonal Antibody for Western Blot, Flow
LZTS2 is a negative regulator of katanin-mediated microtubule severing and release from the centrosome. LZTS2 is required for central spindle formation and the completion of cytokinesis.
Specifications
| PUM2 | |
| Recombinant Monoclonal | |
| 0.55 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q80U58, Q8TB72 | |
| PUM2 | |
| A synthesized peptide derived from human Pumilio 2 (aa 2-46). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB1250 | |
| Unconjugated | |
| PUM2 | |
| 5730503J23Rik; FLJ36528; KIAA0235; MGC138251; MGC138253; mKIAA0235; OTTHUMP00000200539; PUM1; Pum2; PUMH2; pumilio 2; pumilio homolog 2; pumilio RNA binding family member 2; pumilio RNA-binding family member 2; pumilio-2; PUML2; Pumm2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 23369, 298874, 80913 | |
| -20°C | |
| Liquid |