Learn More
Invitrogen™ PUM2 Recombinant Rabbit Monoclonal Antibody (24GB1250)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557657
 View more versions of this product
Product Code. 30275979
  • € 198.00 / 20µL
Description
PUM2 Recombinant Monoclonal Antibody for Western Blot, Flow
LZTS2 is a negative regulator of katanin-mediated microtubule severing and release from the centrosome. LZTS2 is required for central spindle formation and the completion of cytokinesis.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PUM2
Recombinant Monoclonal
0.55 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q80U58, Q8TB72
PUM2
A synthesized peptide derived from human Pumilio 2 (aa 2-46).
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
24GB1250
Unconjugated
PUM2
5730503J23Rik; FLJ36528; KIAA0235; MGC138251; MGC138253; mKIAA0235; OTTHUMP00000200539; PUM1; Pum2; PUMH2; pumilio 2; pumilio homolog 2; pumilio RNA binding family member 2; pumilio RNA-binding family member 2; pumilio-2; PUML2; Pumm2
Rabbit
Affinity chromatography
RUO
23369, 298874, 80913
-20°C