missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PUMA alpha/beta Polyclonal Antibody

PUMA alpha/beta Polyclonal Antibody
Brand: Invitrogen PA5141100
This item is not returnable.
View return policy
Description
PUMA alpha/beta Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
Specifications
| PUMA alpha/beta | |
| Polyclonal | |
| Unconjugated | |
| BBC3 | |
| Bbc3; BCL2 binding component 3; Bcl-2 binding component 3; bcl-2-binding component 3; Bcl-2-binding component 3, isoforms 1/2; Bcl-2-binding component 3, isoforms 3/4; BH3-only; FLJ42994; JFY1; JFY-1; p53 up-regulated modulator of apoptosis; Puma; PUMA/JFY1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 170770, 27113 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.2% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q99ML1, Q9BXH1 | |
| BBC3 | |
| Synthetic peptide within C-terminal residues of PUMA. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction