missing translation for 'onlineSavingsMsg'
Learn More

PUMA Antibody - BSA Free, Novus Biologicals™

Product Code. 18535652 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18535652 0.025 mg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18535652 Supplier Novus Biologicals Supplier No. NBP1766390.025mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 5 publications

PUMA Polyclonal specifically detects PUMA in Human, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PUMA
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2-3 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5-10 μg/mL, Immunocytochemistry/Immunofluorescence 10-20 μg/mL, Immunohistochemistry-Paraffin
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. NP_055232
Gene Alias BCL2 binding component 3, bcl-2-binding component 3, JFY1, JFY-1, p53 up-regulated modulator of apoptosis, PUMAFLJ42994
Gene Symbols BBC3
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 14 amino acids near the amino terminus of human PUMA isoform 1. The immunogen is located within the first 50 amino acids of PUMA. Amino Acid Squence: ARARQEGSSPEPVEG
Molecular Weight of Antigen 23 kDa
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Cellular Markers, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Mitochondrial Markers, Protein Phosphatase, Signal Transduction, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 27113
Test Specificity Human PUMA has 4 isoforms, including isoform 1 (193aa, 21kD), isoform 2 (131aa, 14kD), isoform 3 (101aa, 10kD) and isofrom 4 (261aa, 27kD). This antibody detects human isoform 1, and is predicted to detect mouse and rat PUMA (193aa, 21kD for both of them).
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.