Learn More
Buerkle™ Pump Station for AccuOne/EnergyOne Pump
Shop All Bürkle Products
Click to view available options
Depth (Metric):
50 cm
70 cm
100 cm
Unit Size:
Each
Description
- Pump station accessory only
- Available for three different submersible lengths
Specifications
Specifications
| Depth (Metric) | 70 cm |
| No. per Pack | 1 Ea. |
| Type | Pump Station |
| Applications | Dispensing acids and bases |
| For Use With | AccuOne/EnergyOne Pump |