Learn More
Watson-Marlow Tubing Pumpsil Platinum-Cured Silicone Tubing
Product Code. 30241003
Click to view available options
Diameter (Metric) Inner:
2.4 mm
6.4 mm
8.0 mm
Unit Size:
5m
Product Code. 30241003
Brand: Watson-Marlow Tubing 913.AJ80.016
Only null left
Add to basket
Add to basket
Pumpsil is platinum-cured silicone tubing predominantly used for single-use biopharmaceutical applications, pH control and media feed in fermentation.
  • Developed for the biopharmaceutical industry, Pumpsil is exceptionally pure tubing offering an ultra-smooth bore which helps minimise protein binding and ensure high purity in the finished product.
  • Pumpsil is widely used for single-use biopharm applications, as well as pH control and media feed in fermentation, accurate metering, transfer and filtration. Absolute traceability with laser-etched part number, lot number and use-by-date. Fully documented biocompatibility and comprehensive validation pack.
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Diameter (English) Inner 0.31 in.
Diameter (Metric) Inner 8.0 mm
Material Platinum-cured Silicone
Diameter (English) Outer 0.37 in.
Diameter (Metric) Outer 9.6 mm
Overall Length (English) 16 ft.
Overall Length (Metric) 5 m
Thickness (English) 0.0625 in.
Thickness (Metric) 1.6 mm
Min. Operating Temperature (English) -4°F
Min. Operating Temperature (Metric) -20°C
Color Clear
Hardness 56 (Shore A)
Flexibility Very Flexible
Max. Operating Temperature (English) 176°F
Max. Operating Temperature (Metric) 80°C
Product Type Tubing