missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PURA Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-92651AF594
This item is not returnable.
View return policy
Description
PURA Polyclonal antibody specifically detects PURA in Human, Mouse samples. It is validated for Western Blot
Specifications
| PURA | |
| Polyclonal | |
| Western Blot | |
| PUR1PUR-ALPHA, PURALPHA, purine-rich element binding protein A, Purine-rich single-stranded DNA-binding protein alpha, transcriptional activator protein Pur-alpha | |
| The immunogen this antibody was made to, maps to a region between residue 1 and 50 of human Purine-Rich Element Binding Protein Alpha using the numbering given in entry NP_005850.1 (GeneID 5813). | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 5813 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction